missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SUMO1-Specific Peptidase 1/ SENP1 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6587
This item is not returnable.
View return policy
Description
SENP1 Polyclonal specifically detects SENP1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| SENP1 | |
| Polyclonal | |
| Unconjugated | |
| Q9P0U3 | |
| SENP1 | |
| E. coli-derived recombinant human SUMO1-Specific Peptidase 1/SENP1 Glu419-Leu644 Accession # Q9P0U3 | |
| 100 μg | |
| Primary | |
| Detects human SUMO1-Specific Peptidase 1/SENP1 in direct ELISAs. | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 1-15 ug/mL | |
| EC 3.4.22, EC 3.4.22.-, Sentrin/SUMO-specific protease SENP1, sentrin-specific protease 1, SUMO1/sentrin specific peptidase 1, SUMO1/sentrin specific protease 1, SuPr-2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 29843 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction