missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sulfatase Modifying Factor 2/ SUMF2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2779
This item is not returnable.
View return policy
Description
Sulfatase Modifying Factor 2/SUMF2 Polyclonal specifically detects Sulfatase Modifying Factor 2/SUMF2 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Sulfatase Modifying Factor 2/SUMF2 | |
| Polyclonal | |
| Unconjugated | |
| Q8NBJ7 | |
| SUMF2 | |
| Mouse myeloma cell line NS0-derived recombinant human Sulfatase Modifying Factor 2/SUMF2 Gln26-Leu301 Accession # Q8NBJ7 | |
| 100 μg | |
| Primary | |
| Detects human Sulfatase Modifying Factor 2/SUMF2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 45% cross-reactivity with recombinant mouse SUMF2 is observed and less than 1% cross-reactivity with recombinant human SUMF1 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| C-alpha-formyglycine-generating enzyme 2, C-alpha-formylglycine-generating enzyme 2, DKFZp566I1024, DKFZp686I1024, DKFZp686L17160, DKFZp781L1035, FGE2, MGC99485, paralog of the formylglycine-generating enzyme, pFGE, sulfatase modifying factor 2, sulfatase-modifying factor 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 25870 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction