missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SUGP1 (aa 12-118) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (86%), Rat (86%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52130 (PA5-52130. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
SF4 is a member of the SURP family of splicing factors.
Specifications
Specifications
| Accession Number | Q8IWZ8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 57794 |
| Name | Human SUGP1 (aa 12-118) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 5730496N02Rik; DKFZp434E2216; F23858; RBP; RNA-binding protein RBP; SF4; Splicing factor 4; SUGP1; SURP and G patch domain containing 1; SURP and G-patch domain containing 1; SURP and G-patch domain-containing protein 1 |
| Common Name | SUGP1 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction