missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STRO-1 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB1038U-100UG
This item is not returnable.
View return policy
Description
STRO-1 Monoclonal specifically detects STRO-1 in Human samples. It is validated for Flow Cytometry.
Specifications
| STRO-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| STRO1 | |
| Human CD34 + bone marrow cells | |
| RUO | |
| Detects human STRO-1. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| STRO-1 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction