missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STRBP (aa 270-418) Control Fragment Recombinant Protein

Product Code. 30202715
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30202715

Brand: Invitrogen™ RP102301

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (99%), Rat (99%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54256 (PA5-54256. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Involved in spermatogenesis and sperm function. Plays a role in regulation of cell growth. Binds to double-stranded DNA and RNA. Binds most efficiently to poly(I:C) RNA than to poly(dI:dC) DNA. Binds also to single-stranded poly(G) RNA. Binds non-specifically to the mRNA PRM1 3'-UTR and adenovirus VA RNA.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96SI9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 55342
Name Human STRBP (aa 270-418) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 6430510M02Rik; 74 kDa double-stranded RNA-binding protein; AI852513; C230082I21Rik; C86322; DKFZp434N214; double-stranded RNA-binding protein p74; epididymis luminal protein 162; FLJ11307; FLJ14223; FLJ14984; HEL162; ILF3L; MGC21529; MGC3405; p74; spermatid perinuclear RNA binding protein; spermatid perinuclear RNA-binding protein; SPNR; Strbp
Common Name STRBP
Gene Symbol STRBP
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence CLASGILLPGGPGLHDPCERDPTDALSYMTIQQKEDITHSAQHALRLSAFGQIYKVLEMDPLPSSKPFQKYSWSVTDKEGAGSSALKRPFEDGLGDDKDPNKKMKRNLRKILDSKAIDLMNALMRLNQIRPGLQYKLLSQSGPVHAPVF
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.