missing translation for 'onlineSavingsMsg'
Learn More

Human Stomach Total RNA for QuantiGene Assays

Product Code. 32096702
This item is not returnable. View return policy
Product Code. 32096702 Supplier Invitrogen Supplier No. QS0628
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Species Human
Sample Type RNA
For Use With (Application) Nucleic Acid Quantitation
Product Type Total RNA
Quantity 10 μg
Sample Source Pooled, Normal Tissue
Shipping Condition Dry Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.