missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STK39 (aa 507-536) Control Fragment Recombinant Protein

Product Code. 30195823
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30195823 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30195823 Supplier Invitrogen™ Supplier No. RP104045

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111538 (PA5-111538. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

STK39 (STLK3) is involved in the cellular stress response pathway. STK39 is activated in response to hypotonic stress leading to phosphorylation of several cation-chloride-coupled co-transporters. STK39 activates the p38 MAP kinase pathway and its interaction with p38 decreases during cellular stress. STK39 acts as an intermediate in the response to cellular stress. STK39 is also an independent risk factor for hypertension in men and its intragenic SNPs can interact and function in the control of blood pressure.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UEW8
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 27347
Name Human STK39 (aa 507-536) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW227544; AW556857; DCHT; DKFZp686K05124; kidney-specific SPAK; pancreatic serine/threonine-protein kinase; Pask; proline-alanine-rich STE20-related kinase; PS/TK; PSTK1; RF005; Rnl5; serine threonine kinase 39; serine threonine kinase 39 (STE20/SPS1 homolog, yeast); serine/threonine kinase 39; serine/threonine kinase 39, STE20/SPS1 homolog; serine/threonine-protein kinase 39; small intestine SPAK-like kinase; Spak; Ste-20 related kinase; STE20/SPS1 homolog; STE20/SPS1-related proline-alanine-rich protein kinase; Ste20-like protein kinase; ste-20-related kinase; Stk39
Common Name STK39
Gene Symbol STK39
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IVDDPKALKTLTFKLASGCDGSEIPDEVKL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.