missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human STK11IP (aa 631-707) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (66%), Rat (66%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-111369 (PA5-111369. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May regulate STK11/LKB1 function by controlling its subcellular localization.
Specifications
Specifications
| Accession Number | Q8N1F8 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 114790 |
| Name | Human STK11IP (aa 631-707) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 1200014D22Rik; BB131189; KIAA1898; LIP1; LKB1-interacting protein 1; LKB1IP; serine/threonine kinase 11 interacting protein; serine/threonine kinase 11-interacting protein; serine/threonine-protein kinase 11-interacting protein; STK11 interacting protein; STK11IP; STK11IP1 |
| Common Name | STK11IP |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction