missing translation for 'onlineSavingsMsg'
Learn More

Lonza™ Human Stem Cell Nucleofector™ Kit 1

Product Code. 11566459
Click to view available options
Quantity:
2.25 mL Nucleofector™ Solution for Human Stem Cells, 0.5 mL Supplement 1, 30 μg pmaxGFP™ Vector (0.5 μg/μL in 10 mM Tris pH 8.0), 25 x Aluminum cuvettes (100 μL), 25 x Single-use pipettes
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11566459

Brand: Lonza™ VPH5012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For efficient transfection of human stem cells in the Nucleofector™ I/II/2b Device.

  • High transfection efficiencies for primary cells.
  • Cells maintain functionality post transfection.
  • Applications:
    • Transfection of cell numbers in the range of 2 × 10to 2 × 10cells.
    • Low-throughput transfection in a single cuvette.
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Vector pmaxGFP™
For Use With (Equipment) Nucleofector™ I/II/2b Device
For Use With (Application) Efficient transfection of human stem cells
Includes Nucleofector™ Solution for Human Stem Cells, Supplement 1, pmaxGFP™ Vector (0.5 μg/μL in 10 mM Tris pH 8.0), Aluminum cuvettes, Single-use pipettes
No. of Reactions 25
pH 8
Quantity 2.25 mL Nucleofector™ Solution for Human Stem Cells, 0.5 mL Supplement 1, 30 μg pmaxGFP™ Vector (0.5 μg/μL in 10 mM Tris pH 8.0), 25 x Aluminum cuvettes (100 μL), 25 x Single-use pipettes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.