missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STEAP1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5587-SP
This item is not returnable.
View return policy
Description
STEAP1 Polyclonal specifically detects STEAP1 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| STEAP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.16.1.-, PRSS24, PRSS24MGC19484, six transmembrane epithelial antigen of the prostate, six transmembrane epithelial antigen of the prostate 1, Six-transmembrane epithelial antigen of prostate 1, STEAPmetalloreductase STEAP1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 26872 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL | |
| Q9UHE8 | |
| STEAP1 | |
| E. coli-derived recombinant human STEAP1 Met1-His72 Accession # Q9UHE8 | |
| 25 μg | |
| Primary | |
| Detects human STEAP1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction