missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human STCH (aa 210-339) Control Fragment Recombinant Protein

Product Code. 30199734
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30199734

Brand: Invitrogen™ RP90338

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (85%), Rat (85%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53237 (PA5-53237. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

STCH encodes a protein that is a member of the heat shock protein 70 family and is found associated with microsomes. Members of this protein family play a role in the processing of cytosolic and secretory proteins, as well as in the removal of denatured or incorrectly-folded proteins. The encoded protein contains an ATPase domain and has been shown to associate with a ubiquitin-like protein.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P48723
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6782
Name Human STCH (aa 210-339) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1600002I10Rik; AV006182; B230217N24Rik; Heat shock 70 kDa protein 13; heat shock protein 13; heat shock protein 70 family, member 13; heat shock protein 70 kDa family member 13; heat shock protein 70 kDa family, member 13; heat shock protein family A (Hsp70) member 13; Hspa13; microsomal stress 70 protein ATPase core; microsomal stress-70 protein ATPase core; STCH; stress 70 protein chaperone microsome-associated 60 kDa protein; stress 70 protein chaperone microsome-associated 60 kD human homolog; stress 70 protein chaperone, microsome-associated, 60 kD; stress 70 protein chaperone, microsome-associated, 60 kD human homolog; stress 70 protein chaperone, microsome-associated, 60 kDa; stress 70 protein chaperone, microsome-associated, human homolog; stress-70 protein chaperone microsome-associated 60 kDa protein; testis secretory sperm-binding protein Li 199 A
Common Name STCH
Gene Symbol Hspa13
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AYGLHKADVFHVLVIDLGGGTLDVSLLNKQGGMFLTRAMSGNNKLGGQDFNQRLLQYLYKQIYQTYGFVPSRKEEIHRLRQAVEMVKLNLTLHQSAQLSVLLTVEEQDRKEPHSSDTELPKDKLSSADDH
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.