missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STAT2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB16662-100
This item is not returnable.
View return policy
Description
STAT2 Monoclonal specifically detects STAT2 in Human samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| STAT2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| interferon alpha induced transcriptional activator, ISGF-3, MGC59816, p113, signal transducer and activator of transcription 2, signal transducer and activator of transcription 2, 113kD, signal transducer and activator of transcription 2, 113kDa, STAT113 | |
| E. coli-derived recombinant human STAT2 Gln679-Phe851 Accession # P52630 | |
| 100 μg | |
| Primary | |
| Detects human STAT2 in direct ELISAs and Western blots. | |
| Human | |
| IgG2a |
| Western Blot, KnockDown | |
| 967425 | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Knockout Validated | |
| P52630 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6773 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction