missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human STAT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB14901
This item is not returnable.
View return policy
Description
STAT1 Monoclonal specifically detects STAT1 in Human samples. It is validated for Western Blot, Knockout Validated.
Specifications
| STAT1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Knockout Validated | |
| P42224 | |
| STAT1 | |
| E.coli-derived recombinant human STAT1 Ala687-Val750 Accession # P42224 | |
| 100 μg | |
| Primary | |
| Detects endogenous human STAT1 in Western blots. | |
| Human | |
| Purified |
| Western Blot, KnockDown | |
| 655210 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CANDF7, DKFZp686B04100, ISGF-3, ISGF-3,91kD, signal transducer and activator of transcription 1, 91kDa, signal transducer and activator of transcription-1, STAT91 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6772 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction