missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Stabilin-2 Biotinylated Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems BAF3645
This item is not returnable.
View return policy
Description
Stabilin-2 Polyclonal specifically detects Stabilin-2 in Human samples. It is validated for Western Blot.
Specifications
| Stabilin-2 | |
| Polyclonal | |
| Biotin | |
| Q8WWQ8 | |
| STAB2 | |
| E. coli-derived recombinant human Stabilin-2 Gly2198-Val2295 Accession # Q8WWQ8 | |
| 50 μg | |
| Primary | |
| Detects human Stabilin-2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human Stabilin-1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CD44-like precursor FELL, DKFZP434E0321, FAS1 EGF-like and X-link domain containing adhesion molecule-2, FAS1 EGF-like and X-link domain-containing adhesion molecule 2, fasciclin egf-like, laminin-type egf-like, and link domain-containing scavengerreceptor-2, Fasciclin, EGF-like, laminin-type EGF-like and link domain-containing scavengerreceptor 2, FEEL2, FEEL-2, FELE-2, FELL, FELL-2, FEX2, HARE, HAREFELL2, hepatic hyaluronan clearance receptor, Hyaluronan receptor for endocytosis, hyaluronic acid receptor for endocytosis, STAB2, STAB-2, stabilin 2, stabilin-2, Stabilin2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 55576 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction