missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human ST6GAL2 (aa 40-167) Control Fragment Recombinant Protein

Product Code. 30211459
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30211459

Brand: Invitrogen™ RP90309

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (59%), Rat (59%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-53169 (PA5-53169. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Transfers sialic acid from the donor of substrate CMP-sialic acid to galactose containing acceptor substrates. Has alpha-2,6-sialyltransferase activity toward oligosaccharides that have the Gal-beta-1,4-GlcNAc sequence at the non-reducing end of their carbohydrate groups, but it has weak or no activities toward glycoproteins and glycolipids.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96JF0
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84620
Name Human ST6GAL2 (aa 40-167) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias alpha 2,6 sialyltransferase; alpha 2,6-ST 2; alpha-2,6-sialyltransferase; alpha-2,6-sialyltransferase ST6Gal II; beta galactoside alpha 2,6 sialyltransferase 2; beta-galactosamide alpha-2,6-sialyltransferase II; beta-galactoside alpha-2,6-sialyltransferase 2; beta-galactoside alpha-2,6-sialyltransferase II; C230064G14Rik; CMP-N-acetylneuraminate-beta-galactosamide-alpha-2,6-sialyltransferase 2; hST6Gal II; Kiaa1877; LOW QUALITY PROTEIN: beta-galactoside alpha-2,6-sialyltransferase 2; mKIAA1877; sialyltransferase; sialyltransferase 2; sialyltransferase 2 (monosialoganglioside sialyltransferase); SIAT2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2; ST6 beta-galactosamide alpha-2,6-sialyltranferase 2 A; ST6 beta-galactoside alpha-2,6-sialyltransferase 2; ST6Gal II; st6gal2; st6gal2a; ST6GalII; ST6Gal-II
Common Name ST6GAL2
Gene Symbol ST6GAL2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PSSLSFLETRRLLPVQGKQRAIMGAAHEPSPPGGLDARQALPRAHPAGSFHAGPGDLQKWAQSQDGFEHKEFFSSQVGRKSQSAFYPEDDDYFFAAGQPGWHSHTQGTLGFPSPGEPGPREGAFPAAQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.