missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human ST3GAL5 (aa 236-333) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (93%), Rat (93%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-67026 (PA5-67026. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Ganglioside GM3 is known to participate in the induction of cell differentiation, modulation of cell proliferation, maintenance of fibroblast morphology, signal transduction, and integrin-mediated cell adhesion. The protein encoded by this gene is a type II membrane protein which catalyzes the formation of GM3 using lactosylceramide as the substrate. The encoded protein is a member of glycosyltransferase family 29 and may be localized to the Golgi apparatus. Mutation in this gene has been associated with Amish infantile epilepsy syndrome. Transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
| Accession Number | Q9UNP4 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 8869 |
| Name | Human ST3GAL5 (aa 236-333) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | [a]2; 3 S-T; alpha 2,3-sialyltransferase V; alpha2,3-sialyltransferase; CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; Ganglioside GM3 synthase; GM3 synthase; GM3S; GM3-specific sialytransferase; lactosylceramide alpha-2,3-sialyltransferase; lactosylceramide alpha-2,3-sialyltransferas-like protein; mST3Gal V; SATI; Sialyltransferase 9; sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase); sialyltransferase 9 (CMP-NeuAc:lactosylceramide alpha-2,3-sialyltransferase; GM3 synthase); Siat9; SIATGM3S; ST3 beta-galactoside alpha-2,3-sialyltransferase 5; ST3 beta-galactoside alpha-23-sialyltransferase 5; ST3Gal V; ST3GAL5; ST3GalV; ST3GAL-V; UNQ2510/PRO5998 |
| Common Name | ST3GAL5 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction