missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ST2/IL-33R Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB523-100
This item is not returnable.
View return policy
Description
ST2/IL-33R Monoclonal specifically detects ST2/IL-33R in Human samples. It is validated for Western Blot, Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| ST2/IL-33R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DER4, DER4ST2growth stimulation-expressed, FIT-1, IL-1 R4, IL1R4, IL-1R4, IL-1RL1, IL-33 R, IL33R, IL-33R, interleukin 1 receptor-like 1, interleukin 1 receptor-related protein, interleukin-1 receptor-like 1, Ly84, MGC32623, Protein ST2, ST2L, ST2V, T1, T1homolog of mouse growth stimulation-expressed | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9173 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Neutralization, ELISA | |
| 97203 | |
| Western Blot 1 ug/mL, Neutralization 0.3-1.5 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| BAA02233 | |
| IL1RL1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human ST2/IL-33 R Lys19-Phe328 Accession # BAA02233 | |
| 100 μg | |
| Primary | |
| Detects human ST2/IL-33 R in ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse ST2L, recombinant human (rh) IL-1 RI, rhIL-1 RII, rhIL-1 RA, rhIL-1 alpha , or rhIL-1 beta is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction