missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SSB (aa 15-126) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82583 (PA5-82583. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
La is involved in diverse aspects of RNA metabolism, including binding and protecting 3-prime UUU(OH) elements of newly RNA polymerase III-transcribed RNA, processing 5-prime and 3-prime ends of pre-tRNA precursors, acting as an RNA chaperone, and binding viral RNAs associated with hepatitis C virus. La protein was originally defined by its reactivity with autoantibodies from patients with Sjogren syndrome and systemic lupus erythematosus.
Specifications
Specifications
| Accession Number | P05455 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 6741 |
| Name | Human SSB (aa 15-126) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | autoantigen La; La; la autoantigen; La autoantigen homolog; La protein; la ribonucleoprotein; La ribonucleoprotein domain family, member 3; La/SSB; LARP3; lupus La antigen; lupus La protein; Lupus La protein homolog; Lupus LA protein homolog (LA ribonucleoprotein) (LA autoantigen homolog); lupus La protein-like; mRNA for autoantigen; OTTHUMP00000204777; sjoegren syndrome type B antigen; Sjogren syndrome antigen B; Sjogren syndrome antigen B (autoantigen La); small RNA binding exonuclease protection factor La; Ssb; SS-B; SS-B/La protein |
| Common Name | SSB |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction