missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SRRM1 (aa 850-901) Control Fragment Recombinant Protein

Product Code. 30213440
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30213440

Brand: Invitrogen™ RP106874

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (87%), Rat (87%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66187 (PA5-66187. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Part of pre- and post-splicing multiprotein mRNP complexes. Involved in numerous pre-mRNA processing events. Promotes constitutive and exonic splicing enhancer (ESE)-dependent splicing activation by bridging together sequence-specific (SR family proteins, SFRS4, SFRS5 and TRA2B/SFRS10) and basal snRNP (SNRP70 and SNRPA1) factors of the spliceosome. Stimulates mRNA 3'-end cleavage independently of the formation of an exon junction complex. Binds both pre-mRNA and spliced mRNA 20-25 nt upstream of exon-exon junctions. Binds RNA and DNA with low sequence specificity and has similar preference for either double- or single-stranded nucleic acid substrates.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8IYB3
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10250
Name Human SRRM1 (aa 850-901) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 160-KD; AA407769; LOW QUALITY PROTEIN: serine/arginine repetitive matrix protein 1; plenty-of-prolines 101; plenty-of-prolines-101; Pop101; Ser/Arg-related nuclear matrix protein; serine and arginine repetitive matrix 1; serine/arginine repetitive matrix 1; serine/arginine repetitive matrix protein 1; Srm160; SR-related nuclear matrix protein of 160 kDa; SRRM1; wu:fb34f04; wu:fb38b06; zgc:63802
Common Name SRRM1
Gene Symbol SRRM1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence IAAATTTLAQEEPVAAPEPKKETESEAEDNLDDLEKHLREKALRSMRKAQVS
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.