missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SRPK3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB72301-SP
This item is not returnable.
View return policy
Description
SRPK3/STK23 Monoclonal specifically detects SRPK3/STK23 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| SRPK3/STK23 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| Q9UPE1 | |
| SRPK3 | |
| E. coli-derived recombinant human SRPK3 Arg247-Ser316 Accession # Q9UPE1 | |
| 25 μg | |
| Primary | |
| Detects human SRPK3 in direct ELISAs and Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 722516 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC102944, MSSK1, MSSK-1, MSSK1EC 2.7.11.1, Muscle-specific serine kinase 1, serine arginine rich protein-specific kinase 3, Serine/arginine-rich protein-specific kinase 3, serine/threonine kinase 23, Serine/threonine-protein kinase 23, serine/threonine-protein kinase SRPK3, SFRS protein kinase 3, SR-protein-specific kinase 3, SRSF protein kinase 3, STK23 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26576 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction