missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SRPK2 (aa 274-381) Control Fragment Recombinant Protein

Product Code. 30193881
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30193881

Brand: Invitrogen™ RP91440

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-54272 (PA5-54272. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SRPK2, along with SRPK1, is most likely the cellular protein kinase mediating HBV core protein phosphorylation during viral infection. It therefore represents an important host cell target for therapeutic intervention in HBV infection. Promotes neuronal apoptosis by up-regulating cyclin-D1 (CCND1) expression. This is done by the phosphorylation of SRSF2, leading to the suppression of p53/TP53 phosphorylation thereby relieving the repressive effect of p53/TP53 on cyclin-D1 (CCND1) expression. Phosphorylates ACIN1, and redistributes it from the nuclear speckles to the nucleoplasm, resulting in cyclin A1 but not cyclin A2 up-regulation. Plays an essential role in spliceosomal B complex formation via the phosphorylation of DDX23/PRP28. Plays a negative role in the regulation of HBV replication through a mechanism not involving the phosphorylation of the core protein but by reducing the packaging efficiency of the pregenomic RNA (pgRNA) without affecting the formation of the viral core particles .
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P78362
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6733
Name Human SRPK2 (aa 274-381) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias AW226533; AW492537; AW547358; H_RG152G17.1 A; H_RG152G17.1 b; mSRPK2; serine kinase SRPK2; serine/arginine-rich protein specific kinase 2; serine/arginine-rich protein-specific kinase 2; serine/arginine-rich splicing factor kinase 2; serine/threonine-protein kinase SRPK2; SFRS protein kinase 2; SFRSK2; SR protein kinase 2; SRPK2; SR-protein-specific kinase 2; SRSF protein kinase 2; SRSF protein kinase 2 C-terminal; SRSF protein kinase 2 N-terminal; Wbp6; WUGSC:H_RG152G17.1 A; WW domain binding protein 6
Common Name SRPK2
Gene Symbol SRPK2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence KKQKRQAELLEKRLQEIEELEREAERKIIEENITSAAPSNDQDGEYCPEVKLKTTGLEEAAEAETAKDNGEAEDQEEKEDAEKENIEKDEDDVDQELANIDPTWIESP
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.