missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SRP68 (aa 327-439) Control Fragment Recombinant Protein

Product Code. 30196719
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30196719

Brand: Invitrogen™ RP106650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (94%), Rat (94%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-65959 (PA5-65959. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The signal recognition particle (SRP) is a ribonucleoprotein complex that transports secreted and membrane proteins to the endoplasmic reticulum for processing. The complex includes a 7S RNA and six protein subunits. SRP68 is the 68kDa component of the SRP.The signal recognition particle (SRP) is a ribonucleoprotein complex that transports secreted and membrane proteins to the endoplasmic reticulum for processing. The complex includes a 7S RNA and six protein subunits. This gene encodes the 68kDa component of the SRP. Alternatively spliced transcript variants have been identified, but their biological validity has not been determined. Three related pseudogenes are located within the Smith-Magenis syndrome region on chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9UHB9
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6730
Name Human SRP68 (aa 327-439) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610024I03Rik; 68 kDa signal recognition particle; 68 kDA subunit of signal recognition particle; likely ortholog of H. sapiens signal recognition particle 68 kDa (SRP68); signal recognition particle 68; Signal recognition particle 68 kDa protein; signal recognition particle 68 kD; signal recognition particle 68 kDa; signal recognition particle subunit SRP68; Srp68; zgc:92573
Common Name SRP68
Gene Symbol SRP68
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence AESEETKERLFESMLSECRDAIQVVREELKPDQKQRDYILEGEPGKVSNLQYLHSYLTYIKLSTAIKRNENMAKGLQRALLQQQPEDDSKRSPRPQDLIRLYDIILQNLVELL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.