missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SRGAP3 (aa 905-971) Control Fragment Recombinant Protein
missing translation for 'orderingAttributeHoverText'
Quantity:
100 μL
missing translation for 'unitSize'
100µL
Beschreibung
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-60607 (PA5-60607. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
GTPase-activating protein for RAC1 and perhaps Cdc42, but not for RhoA small GTPase. May attenuate RAC1 signaling in neurons. [UniProt]
Spezifikation
Spezifikation
| Accession Number | O43295 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 9901 |
| Name | Human SRGAP3 (aa 905-971) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | AI452337; ARHGAP14; brain stress early protein Gbi; D130026O08Rik; Gbi; Kiaa0411; KIAA1156; MEGAP; mental disorder-associated GAP; mKIAA0411; RGD1563152; rho GTPase activating protein 14; rho GTPase-activating protein 14; SLIT-ROBO Rho GTPase activating protein 2; SLIT-ROBO Rho GTPase activating protein 3; SLIT-ROBO Rho GTPase-activating protein 3; SRGAP2; srGAP3; WAVE-associated Rac GTPase activating protein; WAVE-associated Rac GTPase-activating protein; WRP |
| Common Name | SRGAP3 |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur