missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SREC-I/SCARF1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2409-SP
This item is not returnable.
View return policy
Description
SREC-I/SCARF1 Polyclonal specifically detects SREC-I/SCARF1 in Human samples. It is validated for Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.
Specifications
| SREC-I/SCARF1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Acetyl LDL receptor, KIAA0149endothelial cells scavenger receptor, MGC47738, SCARF1, scavenger receptor class F, member 1, Scavenger receptor expressed by endothelial cells 1, SREC1, SRECI, SREC-I, SRECscavenger receptor expressed by endothelial cells, SR-F1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8578 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Blockade of Receptor-ligand Interaction, CyTOF-ready | |
| Q14162 | |
| SCARF1 | |
| Mouse myeloma cell line NS0-derived recombinant human SREC-I/SCARF1 Ser20-Thr421 Accession # Q14162 | |
| 25 μg | |
| Primary | |
| Detects SREC-I/SCARF1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) SREC-II is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction