missing translation for 'onlineSavingsMsg'
Learn More

Human SR-BI Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

£170.00 - £369.00

Specifications

Antigen SR-BI
Clone 947007
Dilution Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18792628
GREENER_CHOICE
View Documents
R&D Systems
MAB8114-SP
25 μg
£170.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18192661
View Documents
R&D Systems
MAB8114
100 μg
£369.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SR-BI Monoclonal specifically detects SR-BI in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

SR-BI
Western Blot 2 ug/mL, Immunohistochemistry 5-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
949
Human embryonic kidney cell line HEK293-derived human SR-BI
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human SR-BI in direct ELISAs and Western blots.
947007
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human
CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1, CD36L1scavenger receptor class B type 1, CLA1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, HDLQTL6, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI
SCARB1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.