missing translation for 'onlineSavingsMsg'
Learn More

Human SR-AI/MSR1 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

£181.00 - £450.00

Specifications

Antigen SR-AI/MSR
Clone 351620
Concentration LYOPH
Dilution Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction
Applications Western Blot, Blockade of Receptor-ligand Interaction
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18737158
GREENER_CHOICE
View Documents
R&D Systems
MAB27081-SP
25 μg
£181.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18129020
View Documents
R&D Systems
MAB27081
100 μg
£450.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Human samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.
Specifications

Specifications

SR-AI/MSR
LYOPH
Western Blot, Blockade of Receptor-ligand Interaction
Unconjugated
Mouse
Human
P21757
4481
Mouse myeloma cell line NS0-derived recombinant human SR-AI Lys77-Leu451 Accession # P21757
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human SR-AI in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse SR-AI.
351620
Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI
MSR1
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.