missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SR-AI/MSR Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2708
This item is not returnable.
View return policy
Beskrivelse
SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-reported.
Tekniske data
| SR-AI/MSR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported | |
| P21757 | |
| MSR1 | |
| Mouse myeloma cell line NS0-derived recombinant human SR-AI/MSR Lys77-Leu451 Accession # P21757 | |
| 100 μg | |
| Primary | |
| Detects human SR-AI/MSR in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SR-AI is observed. | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 351615 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, MSR1, phSR1, phSR2, SCARA1, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A, SRAI | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4481 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion