missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPTY2D1 (aa 164-241) Control Fragment Recombinant Protein

Product Code. 30200807
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200807

Brand: Invitrogen™ RP97507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (82%), Rat (82%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58760 (PA5-58760. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Histone chaperone that stabilizes pre-existing histone tetramers and regulates replication-independent histone exchange on chromatin (PubMed:26109053). Required for normal chromatin refolding in the coding region of transcribed genes, and for the suppression of spurious transcription (PubMed:26109053). Binds DNA and histones and promotes nucleosome assembly (in vitro) (PubMed:23378026, PubMed:26109053). Facilitates formation of tetrameric histone complexes containing histone H3 and H4 (PubMed:26109053). Modulates RNA polymerase 1-mediated transcription. Binds DNA, with a preference for branched DNA species, such as Y-form DNA and Holliday junction DNA (PubMed:23378026).
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q68D10
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 144108
Name Human SPTY2D1 (aa 164-241) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias DKFZp686I068; FLJ39441; Protein KU002155; Protein SPT2 homolog; Spt2; SPT2 chromatin protein domain containing 1; SPT2 domain-containing protein 1; SPT2, Suppressor of Ty, domain containing 1; SPTY2D1
Common Name SPTY2D1
Gene Symbol SPTY2D1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PPPMNFTDLLRLAEKKQFEPVEIKVVKKSEERPMTAEELREREFLERKHRRKKLETDGKLPPTVSKKAPSQKESVGTK
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.