missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SPSB3 (aa 124-257) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Description
Highest antigen sequence indentity to the following orthologs: Mouse (97%), Rat (97%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61257 (PA5-61257. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
May be a substrate recognition component of a SCF-like ECS (Elongin BC-CUL2/5-SOCS-box protein) E3 ubiquitin-protein ligase complex which mediates the ubiquitination and subsequent proteasomal degradation of target proteins.
Specifications
Specifications
| Accession Number | Q6PJ21 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 90864 |
| Name | Human SPSB3 (aa 124-257) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 2310012N15Rik; 3300001M01Rik; C16orf31; Kiaa4204; mKIAA4204; splA/ryanodine receptor domain and SOCS box containing 3; SPRY domain-containing SOCS box protein 3; SPRY domain-containing SOCS box protein SSB-3; SPSB3; SSB3; SSB-3; Tce1 |
| Common Name | SPSB3 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction