Learn More
Human SPRY3 Antibody, R&D Systems™
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  R&D Systems AF5735-SP
Product Code. 18776377
  • £146.00 / 25µg
Description
SPRY3 Polyclonal specifically detects SPRY3 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SPRY3
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Antagonist Of FGF Signaling, HSPRY3, Protein Sprouty Homolog 3, sprouty (Drosophila) homolog 2, Sprouty Homolog 3 (Drosophila), spry-3
Sheep
Affinity Purified
RUO
10251
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot
LYOPH
Western Blot 1 ug/mL
O43610
SPRY3
E. coli-derived recombinant human SPRY3 Met1-Glu142 Accession # O43610
25 μg
Primary
Detects endogenous human SPRY3 in Western blots.