missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SPRED1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5067-SP
This item is not returnable.
View return policy
Description
SPRED1 Polyclonal specifically detects SPRED1 in Human samples. It is validated for Western Blot.
Specifications
| SPRED1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 161742 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q7Z699 | |
| SPRED1 | |
| E. coli-derived recombinant human SPRED1 Arg118-Lys324 Accession # Q7Z699 | |
| 25 μg | |
| Primary | |
| Detects endogenous human SPRED1 in Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction