missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPIRE2 (aa 371-448) Control Fragment Recombinant Protein

Product Code. 30181464
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30181464

Brand: Invitrogen™ RP98594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-59561 (PA5-59561. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Acts as an actin nucleation factor, remains associated with the slow-growing pointed end of the new filament (PubMed:21620703). Involved in intracellular vesicle transport along actin fibers, providing a novel link between actin cytoskeleton dynamics and intracellular transport. Required for asymmetric spindle positioning and asymmetric cell division during meiosis (PubMed:21620703). Required for normal formation of the cleavage furrow and for polar body extrusion during female germ cell meiosis (PubMed:21620703). Also acts in the nucleus: together with SPIRE1 and SPIRE2, promotes assembly of nuclear actin filaments in response to DNA damage in order to facilitate movement of chromatin and repair factors after DNA damage (PubMed:26287480). [UniProt]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8WWL2
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 84501
Name Human SPIRE2 (aa 371-448) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BC026502; KIAA1832; Protein spire homolog 2; SPIR2; spir-2; spire actin nucleation factor 2; spire family actin nucleation factor 2; spire homolog 2; spire homolog 2 (Drosophila); spire type actin nucleation factor 2; SPIRE2; spire-type actin nucleation factor 2
Common Name SPIRE2
Gene Symbol SPIRE2
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence SLPCILNACSGDAKSTSCINLSVTDAGGSAQRPRPRVLLKAPTLAEMEEMNTSEEEESPCGEVTLKRDRSFSEHDLAQ
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.