missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sphingosine Kinase 1/SPHK1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5536
This item is not returnable.
View return policy
Description
Sphingosine Kinase 1/SPHK1 Polyclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.
Specifications
| Sphingosine Kinase 1/SPHK1 | |
| Polyclonal | |
| Unconjugated | |
| NP_068807 | |
| SPHK1 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Sphingosine Kinase 1/SPHK1 Asp2-Leu398 Accession # NP_068807 | |
| 100 μg | |
| Primary | |
| Detects human SPHK1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human SPHK2 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| EC 2.7.1.91, SK1, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1, SPK1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8877 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction