missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPERT (aa 179-267) Control Fragment Recombinant Protein

Product Code. 30182497
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30182497

Brand: Invitrogen™ RP97989

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (71%), Rat (71%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-58752 (PA5-58752. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

SPERT (spermatid associated protein), also known as CBY2 (chibby homolog 2) or NURIT, is a novel leucine-zipper protein belonging to the chibby family of proteins. Uniquely Expressed in the spermatid flower-like structure, SPERT interacts with Nek1, a member of the NIMA-family kinase family that is associated centrosomal stability and ciliogenesis. It contains a leucine-zipper motif and two coiled-coil regions, and is transcribed through the elongation stage of the spermatids. SPERT is absent from mature spermatozoa and is thought to be involved in transporting proteins that are to be discarded via the residual bodies.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q8NA61
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 220082
Name Human SPERT (aa 179-267) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700086N05Rik; AI596460; CBY2; chibby family member 2; chibby homolog 2; novel leucine zipper testicular protein; Nurit; Protein chibby homolog 2; protein nurit; spermatid associated; spermatid flower-like structure protein; spermatid-associated protein; Spert; testicular tissue protein Li 182; testis-specific leucine zipper protein nurit
Common Name SPERT
Gene Symbol SPERT
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NKALREENRMLSKENKILQVFWEEHKASLGREESRAPSPLLHKDSASLEVVKKDHVALQVPRGKEDSTLQLLREENRALQQLLEQKQAY
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Vedi altri risultati Mostra meno risultati
Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato