missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human Spectrin alpha-1 (aa 1022-1110) Control Fragment Recombinant Protein

Product Code. 30208024
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30208024

Brand: Invitrogen™ RP94435

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (81%), Rat (81%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-82997 (PA5-82997. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Spectrin is an actin crosslinking and molecular scaffold protein that links the plasma membrane to the actin cytoskeleton, and functions in the determination of cell shape, arrangement of transmembrane proteins, and organization of organelles. It is a tetramer made up of alpha-beta dimers linked in a head-to-head arrangement. This gene is one member of a family of alpha-spectrin genes. The encoded protein is primarily composed of 22 spectrin repeats which are involved in dimer formation. It forms weaker tetramer interactions than non-erythrocytic alpha spectrin, which may increase the plasma membrane elasticity and deformability of red blood cells. Mutations in this gene result in a variety of hereditary red blood cell disorders, including elliptocytosis type 2, pyropoikilocytosis, and spherocytic hemolytic anemia. [provided by RefSeq, Jul 2008]
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P02549
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6708
Name Human Spectrin alpha-1 (aa 1022-1110) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 2610027H02Rik; A2a; AF093576; a--fodrin; AI451697; alpha II spectrin; alpha-fodrin; alpha-I spectrin; alpha-II spectrin; alpha-spectrin 1, erythroid; alpha-spectrin 2; alpha-spectrin 2, brain; brain alpha-spectrin; EIEE5; EL2; elliptocytosis 2; erythroid alpha-spectrin; erythroid spectrin alpha; fodrin alpha chain; ha; hemolytic anemia; HPP; HS3; ihj; inhibitory protein factor; IPF; mutant alpha spectrin; NEAS; neuroscience mutagenesis facility, 4; nmf4; noerythroid alpha-spectrin 2; nonerythroid alpha-spectrin 2; spectrin alpha 1; spectrin alpha chain, brain; spectrin alpha chain, erythrocyte; spectrin alpha chain, erythrocytic 1; Spectrin alpha chain, non-erythrocytic 1; spectrin alpha, erythrocytic 1; spectrin alpha, non-erythrocytic 1; spectrin, alpha, erythrocytic 1; spectrin, alpha, erythrocytic 1 (elliptocytosis 2); spectrin, alpha, non-erythrocytic 1; spectrin, non-erythroid alpha chain; spectrin, non-erythroid alpha subunit; sph; SPH3; spherocytosis; Spna1; Spna-1; Spna2; Spna-2; SPTA; Spta1; SPTA2; SPTAN1
Common Name Spectrin alpha-1
Gene Symbol SPTA1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence HQGIVPAVYVRRLAHDEFPMLPQRRREEPGNITQRQEQIENQYRSLLDRAEERRRRLLQRYNEFLLAYEAGDMLEWIQEKKAENTGVEL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.