Learn More
Invitrogen™ Human CD31/PECAM (Soluble) ELISA Kit
Description
Includes
Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sPECAM-1
HRP-Conjugate anti-human sPECAM-1 monoclonal antibody
Human sPECAM-1 Standard, 60ng/mL
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films
The Human Soluble CD31/Platelet Endothelial Cell Adhesion Molecule(Hu sCD31/PECAM) ELISA quantitates Hu sCD31/PECAM in human serum, plasma, cell culture supernatants, urine, or other body fluids. The assay will exclusively recognize both natural and recombinant Hu sCD31/PECAM. Principle of the method The Human sCD31/PECAM solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | P16284 |
| Assay Range | 0.47 to 30.0 ng/mL |
| Assay Sensitivity | 0.06 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant, Urine |
| Target Species | Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 5175 |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.