missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SPARC/Osteonectin Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems BAF941
This item is not returnable.
View return policy
Description
SPARC Polyclonal specifically detects SPARC in Human samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry.
Specifications
| SPARC | |
| Polyclonal | |
| Biotin | |
| P09486 | |
| SPARC | |
| Mouse myeloma cell line NS0-derived recombinant human SPARC/Osteonectin Ala18-Ile303 Accession # P09486 | |
| 50 μg | |
| Primary | |
| Detects human SPARC in Western blots. In Western blots, less than 1% cross-reactivity with recombinant mouse SPARC is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells | |
| Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6678 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction