missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPANXD (aa 62-86) Control Fragment Recombinant Protein

Product Code. 30212566
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30212566

Brand: Invitrogen™ RP108618

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (44%), Rat (44%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-84932 (PA5-84932. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Temporally regulated transcription and translation of several testis-specific genes is required to initiate the series of molecular and morphological changes in the male germ cell lineage necessary for the formation of mature spermatozoa. This gene is a member of the SPANX family of cancer/testis-associated genes, which are located in a cluster on chromosome X. The SPANX genes encode differentially expressed testis-specific proteins that localize to various subcellular compartments. This particular gene encodes a sperm protein that is associated with the nucleus but, although a role in spermatogenesis is suggested, the specific function of this family member has not yet been determined. Polymorphisms in this gene may be associated with prostate cancer susceptibility. [provided by RefSeq, Apr 2014].
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXN6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 64648
Name Human SPANXD (aa 62-86) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias cancer/testis antigen 11.4; cancer/testis antigen family 11, member 4; CT11.4; dJ171K16.1; Nuclear-associated protein SPAN-Xd; nuclear-associated protein SPAN-Xe; SPANX family member D; SPANX family, member D; SPANX family, member E; SPANX-C; SPANXD; SPANX-D; SPANXE; SPANX-E; Sperm protein associated with the nucleus on the x chromosome D; sperm protein associated with the nucleus, x chromosome, family member D; sperm protein associated with the nucleus, x chromosome, family member E
Common Name SPANXD
Gene Symbol SPANXD
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence TSPEELVNDHARKNRINPLQMEEEE
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.