missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human SPAG4 (aa 191-279) Control Fragment Recombinant Protein

Product Code. 30200638
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
This item is not returnable. View return policy

Product Code. 30200638

Brand: Invitrogen™ RP100903

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-61732 (PA5-61732. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NPE6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 6676
Name Human SPAG4 (aa 191-279) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias 1700041K21Rik; acrosomal protein ACR55; cancer/testis antigen 127; CT127; mKIAA4118; MNCb-0953; outer dense fiber-associated protein SPAG4; Sad1 and UNC84 domain containing 4; SPAG4; sperm antigen 4; sperm associated antigen 4; sperm tail protein; sperm-associated antigen 4 protein; SUN domain-containing protein 4; SUN4
Common Name SPAG4
Gene Symbol SPAG4
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence PLENEPKEMLTLSEYHERVRSQGQQLQQLQAELDKLHKEVSTVRAANSERVAKLVFQRLNEDFVRKPDYALSSVGASIDLQKTSHDYAD
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.