missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SP-D Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB19201-SP
This item is not returnable.
View return policy
Description
SP-D Monoclonal specifically detects SP-D in Human samples. It is validated for ELISA.
Specifications
| SP-D | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| COLEC7collectin-7, collectin 7, Collectin-7, Lung surfactant protein D, PSPD, PSP-D, SFTP4, SFTP4pulmonary surfactant-associated protein D, SFTPD, SPD, SP-Dpulmonary surfactant apoprotein, surfactant protein D, surfactant, pulmonary-associated protein D, surfactant-associated protein, pulmonary 4 | |
| Mouse myeloma cell line NS0-derived recombinant human SP-D, Ala21-Phe375, Accession # P35247.2 | |
| 25 μg | |
| 6441 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG2b |
| ELISA | |
| 292215 | |
| ELISA | |
| P35247.2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human SP-D in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction