missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human SP-B/Surfactant Protein B (AAH32785) VersaClone cDNA

Product Code. 18764728
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18764728 10 μg 10µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18764728 Supplier R&D Systems™ Supplier No. RDC1999

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

R&D Systems™ Human SP-B/Surfactant Protein B (AAH32785) VersaClone cDNA is an amphipathic pulmonary-associated surfactant protein essential for lung function and homeostasis after birth.

The Human SP-B/Surfactant Protein B (AAH32785) VersaClone cDNA clone contains the complete ORF for SP-B/Surfactant Protein B, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The SP-B/Surfactant Protein B insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY

Specifications

Gene hSFTPB
Accession Number AAH32785
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl
Insert Type 1159bp
pH 8.5
Gene ID (Entrez) 6439
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Human
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.