missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX3 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF2569-SP
This item is not returnable.
View return policy
Description
SOX3 Polyclonal specifically detects SOX3 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| SOX3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| GHDX, MRGH, panhypopituitarism, PHP, PHPX, SOXB, SRY (sex determining region Y)-box 3, transcription factor SOX-3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6658 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| P41225 | |
| SOX3 | |
| E. coli-derived recombinant human SOX3 Met1-Leu230 Accession # P41225 | |
| 25 μg | |
| Primary | |
| Detects human SOX3 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 1% cross-reactivity with recombinant human (rh) SOX2, rhSOX7, rhSOX10, and rhSOX17 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction