missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX17 Antibody, R&D Systems™
Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB19241
This item is not returnable.
View return policy
Description
SOX17 Monoclonal specifically detects SOX17 in Human samples. It is validated for Immunocytochemistry.
Specifications
| SOX17 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| Q9H6I2 | |
| SOX17 | |
| E. coli-derived recombinant human SOX17 Asp177-Val414 Accession # Q9H6I2 | |
| 100 μg | |
| Primary | |
| Detects human SOX17 in direct ELISAs. | |
| Human | |
| Purified |
| Immunocytochemistry | |
| 614013 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ22252, SRY (sex determining region Y)-box 17, SRY-related HMG-box transcription factor SOX17, transcription factor SOX-17, VUR3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 64321 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction