missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOX15 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4070-SP
This item is not returnable.
View return policy
Description
SOX15 Polyclonal specifically detects SOX15 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| SOX15 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| protein SOX-15, SOX20, SOX20Protein SOX-20, SOX26, SOX26SOX12, SOX27, SOX27Protein SOX-12, SRY (sex determining region Y)-box 15, SRY (sex determining region Y)-box 20 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 6665 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| O60248 | |
| SOX15 | |
| E. coli-derived recombinant human SOX15 Met1-Leu233 Accession # O60248 | |
| 25 μg | |
| Primary | |
| Detects human SOX15 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human (rh) SOX3, rhSOX14, and rhSOX21 is observed. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction