missing translation for 'onlineSavingsMsg'
Learn More

Human SOX14 Antibody, R&D Systems™

Product Code. 30689910
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30689910 25 μg 25µg
30689863 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30689910 Supplier R&D Systems Supplier No. MAB11693SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

SOX14 Monoclonal antibody specifically detects SOX14 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen SOX14
Applications Immunohistochemistry
Classification Monoclonal
Clone 1100814
Conjugate Unconjugated
Dilution Immunohistochemistry 3-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene Accession No. O95416
Gene Alias HMG box transcription factor SOX-14, MGC119898, MGC119899, SOX28, SRY (sex determining region Y)-box 14, SRY-box 14, transcription factor SOX-14
Host Species Mouse
Immunogen Synthetic Peptide, Accession # O95416
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8403
Reconstitution Reconstitute lyophilized material at 0.2 mg/ml in sterile PBS. For liquid material, refer to CoA for concentration.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.