missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human SOST/Sclerostin Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 7 publications
Brand: R&D Systems MAB1406-100
This item is not returnable.
View return policy
Description
SOST/Sclerostin Monoclonal specifically detects SOST/Sclerostin in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
| SOST/Sclerostin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Q9BQB4 | |
| SOST | |
| Mouse myeloma cell line NS0-derived recombinant human SOST/Sclerostin Gln24-Tyr213 Accession # Q9BQB4 | |
| 100 μg | |
| Primary | |
| Detects human SOST/Sclerostin in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse SOST is observed. | |
| Human | |
| Supernatant |
| Western Blot, ELISA | |
| 220902 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| sclerostin, VBCHsclerosteosis | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 50964 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction