missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sortilin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB31541-100
This item is not returnable.
View return policy
Description
Sortilin Monoclonal specifically detects Sortilin in Human samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Sortilin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 100 kDa NT receptor, Glycoprotein 95, Gp95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, Ntr3, SORT1, sortilin, sortilin 1 | |
| Mouse myeloma cell line NS0-derived recombinant human Sortilin Ser78-Asn755 Accession # Q99523 | |
| 100 μg | |
| 6272 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| CyTOF, Flow Cytometry | |
| 334703 | |
| Flow Cytometry 0.25μg/106 cells, CyTOF-ready | |
| Q99523 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human Sortilin in direct ELISAs. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction