Learn More
ACROBiosystems Human Sonic Hedgehog / Shh Protein, His Tag
Description
Sonic hedgehog protein (SHH) is also known as HHG-1, which belongs to the hedgehog family. SHH is expressed in fetal intestine, liver, lung, and kidney and not expressed in adult tissues. Sonic hedgehog protein can be cleavage into sonic hedgehog protein N-product and sonic hedgehog protein C-product by autolysis. SHH binds to the patched (PTC) receptor, which functions in association with smoothened (SMO), to activate the transcription of target genes. In the absence of SHH, PTC represses the constitutive signaling activity of SMO. Also regulates another target, the gli oncogene. Intercellular signal essential for a variety of patterning events during development. The C-terminal domain displays an autoproteolysis activity and a cholesterol transferase activity. The N-product is the active species in both local and long-range signaling, whereas the C-product has no signaling activity.
Specifications
Specifications
| Accession Number | NP_000184 |
| Molecular Weight (g/mol) | 20.4 kDa |
| Quantity | 1 mg |
| Source | Human Sonic Hedgehog, His Tag (SH7-H5229) is expressed from human 293 cells (HEK293). It contains AA Cys 24 - Gly 197 (Accession # NP_000184). |
| Immunogen | Cys 24 - Gly 197 |
| Storage Requirements | -20°C |
| Gene Alias | SHH,HHG-1,Sonic hedgehog,HLP3,HPE3,SMMCI,TPT,TPTPS |
| Common Name | Sonic Hedgehog |
| Conjugate | Unconjugated |
| Species | Human |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.