Learn More
R&D Systems™ Human Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit
Product Code. 18752404
Click to view available options
Unit Size:
1 set
Product Code. 18752404
Brand: R&D Systems™ DSHH00
Human Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
The Human Sonic Hedgehog/Shh N-Terminus Quantikine ELISA Kit quantifies human Sonic Hedgehog/Shh in cell culture supernates (34 ul), serum (34 ul), heparin plasma (34 ul).
TRUSTED_SUSTAINABILITY
Specifications
Sample Type Cell Culture Supernatant, Plasma (Heparin), Serum