missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Sonic Hedgehog/Shh Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £418.00
Spécification
| Antigen | Sonic Hedgehog/Shh |
|---|---|
| Clone | 605022 |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
18711677
![]() |
R&D Systems
MAB109391-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18700307
![]() |
R&D Systems
MAB109391-100 |
100 μg |
£418.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Sonic Hedgehog/Shh Monoclonal antibody specifically detects Sonic Hedgehog/Shh in Human samples. It is validated for nanSpécification
| Sonic Hedgehog/Shh | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| HHG1, HHG-1, HLP3, HPE3, MCOPCB5, MCOPCB5sonic hedgehog (Drosophila) homolog, Shh, ShhNC, SMMCI, SMMCIsonic hedgehog homolog (Drosophila), sonic hedgehog, sonic hedgehog homolog, sonic hedgehog protein, TPT, TPTPS | |
| E. coli-derived human Sonic Hedgehog/Shh protein, Cys24-Gly197, Accession # NP_000184.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 605022 | |
| Unconjugated | |
| Mouse | |
| Human | |
| NP_000184.1 | |
| 6469 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit